GC Analysis of Natural Gas (GPA Method 2261)
Conditions | |
---|---|
column | GPA 2261 3-column set (13993-U) |
oven | 100 °C |
carrier gas | helium, 30 mL/min |
injection | 0.5 mLsample loop |
Description | |
---|---|
Analysis Note | Packed columns continue to be used in multi-column, multi-valve configurations for the determination of BTU content in natural gas and natural gas liquids as described in Gas Processors Association (GPA) Methods 2261 and 2177, plus American Society for Testing and Materials (ASTM) Test Method 2597. This chromatogram is of GPA Method 2261, and uses three packed columns in a multi-valve configuration. Supelco is the world leader in packed GC column technology, with an extensive library of phases and supports. |
Categories | Analytical Chromatography, GC Applications, Volatiles, Gases, Hydrocarbons |
Featured Industry | Petroleum |
Other Notes | Complementary products: Syringes Vials Gas Purification and Gas Management Inlet Liners, Glass Wool, and Specialized Hand Tools Septa and Specialized Hand Tools Column Ferrules, Nuts, and Specialized Hand Tools |
suitability | application for GC |
Materials
Sorry, an unexpected error has occurred
Network error: Failed to fetch