MilliporeSigma
UHPLC Gradient Analysis of Boswellia Herbal Supplement on Titan C18
UHPLC Gradient Analysis of Boswellia Herbal Supplement on Titan C18 application for UHPLC
Materials
mobile phase component
Product No.
Description
Pricing
Phosphoric acid
≥85 wt. % in H2O, ≥99.999% trace metals basis
CONDITIONS
extraction
1. Weigh 100 mg Bosewellia Herbal Supplement     2. Add 10 mL ethanol     3. Sonicate 30 min at 40°C     4. Dilute 100 μL extract, 400 μL ethanol, 500 μL water
column
Titan C18, 10 cm x 2.1 mm I.D., 1.9 μm particles (577124-U)
mobile phase
[A] 0.1% phosphoric acid; [B]acetonitrile
gradient
40% B for 0.5 min; to 100% B in 2 min; held at 100% B for 10 min
flow rate
0.4 mL/min
pressure
4050 psi (279 bar)
column temp.
35 °C
detector
UV, 210 nm
injection
2 μL
sample
Bosewellia Extract in 50:50, water:ethanol
Description
Legal Information
null