MilliporeSigma
HPLC Analysis of Fat Soluble Vitamins by Normal Phase Chromatography on Ascentis® Si (25 cm Column)
HPLC Analysis of Fat Soluble Vitamins by Normal Phase Chromatography on Ascentis® Si (25 cm Column) application for HPLC
Materials
Analytical column
Product No.
Description
Pricing
Ascentis® Si HPLC Column
5 μm particle size, L × I.D. 15 cm × 4.6 mm
standard
Product No.
Description
Pricing
Cholecalciferol (D3)
analytical standard
Menadione (K3)
analytical standard
(±)-α-Tocopherol
analytical standard
(+)-γ-Tocopherol
analytical standard
CONDITIONS
column
Ascentis Si, 15 cm x 4.6 mm I.D., 5 μm particles (581512-U)
mobile phase
[A] hexane; [B] ethyl acetate
gradient
10 to 30% B in 10 min; held at 30% B for 2 min
flow rate
1.0 mL/min
column temp.
30 °C
detector
UV, 290 nm
injection
10 μL
Description
Analysis Note
Ascentis silica is used in the analysis of fat-soluble vitamins using a normal phase gradient. These include vitamin K 3 (menadione), vitamin E (alpha and gamma tocopherol), and vitamin D3 (cholecalciferol).
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany