MilliporeSigma
OSHA Method 42/47 and ASTM® D5836: HPLC Analysis of Isocyanates on SUPELCOSIL LC-8 after Collection/Desorption using ORBO-80
OSHA Method 42/47 and ASTM® D5836: HPLC Analysis of Isocyanates on SUPELCOSIL™ LC-8 after Collection/Desorption using ORBO™-80 application for air monitoring, application for HPLC
Materials
Analytical column
Product No.
Description
Pricing
SUPELCOSIL LC-8 HPLC Column
5 μm particle size, L × I.D. 25 cm × 4.6 mm
related product
Product No.
Description
Pricing
ORBO 80 Isocyanate Filter
with cassettes: no, filter O.D. 37 mm, pkg of 25 ea
CONDITIONS
sample/matrix
calibration standard: isocyanate derivatives, 5 μg/mL in acetonitrile blank: 1-(2-pyridyl)piperazine (1-2PP) coated glass fiber filter desorbed in acetonitrile/DMSO, 9:1
adsorbent tube
ORBO-80 (20811)
column
SUPELCOSIL LC-8, 25 cm × 4.6 mm I.D., 5 μm particles (58297)
mobile phase
0.05 M ammonium acetate in water:acetonitrile, 70:30 (pH 6-6.2 with acetic acid)
flow rate
linear gradient, 0.7 mL/min to 2 mL/min in 15 min
detector
UV, 254 nm
injection
10 μL
Description
Legal Information
ASTM is a registered trademark of American Society for Testing and Materials
ORBO is a trademark of Sigma-Aldrich Co. LLC
SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC