HPLC Analysis of Lysozyme, Myoglobin and Impurities on Discovery® BIO Wide Pore C5
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
Discovery BIO Wide Pore C5, 15cm × 4.6mm I.D., 5μm particles (568422-U)
mobile phase
[A] (75:25) 0.1% TFA in water:0.1% TFA in acetonitrile; [B] (25:75) 0.1% TFA in water:0.1% TFA in acetonitrile
gradient
0-100% B in 25 min
flow rate
1.0 mL/min
column temp.
ambient
detector
UV, 220 nm
injection
12 μL in 0.1% TFA
Description
Legal Information
Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany