MilliporeSigma
HPLC Analysis of Vitamins, Fat Soluble (D2 and D3), on Discovery® HS C18
HPLC Analysis of Vitamins, Fat Soluble (D2 and D3), on Discovery® HS C18 application for HPLC
Materials
Analytical column
Product No.
Description
Pricing
Discovery® HS C18 HPLC Column
3 μm particle size, L × I.D. 15 cm × 4.6 mm
standard
Product No.
Description
Pricing
Cholecalciferol (D3)
analytical standard
Ergocalciferol (D2)
analytical standard
CONDITIONS
column
Discovery HS C18, 15 cm × 4.6 mm I.D., 3 μm particles (569252-U)
mobile phase
100% acetonitrile
flow rate
0.8 mL/min
column temp.
30 °C
detector
UV, 290 nm
injection
10 μL
sample
50 μg/mL each analyte
Description
Legal Information
Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany