MilliporeSigma
HPLC Analysis of Lansoprazole Enantiomers on Astec® CYCLOBOND I 2000 DMP
HPLC Analysis of Lansoprazole Enantiomers on Astec® CYCLOBOND I 2000 DMP application for HPLC
Materials
Analytical column
Product No.
Description
Pricing
Astec® CYCLOBOND I 2000 DMP Chiral HPLC Column
5 μm particle size, L × I.D. 25 cm × 4.6 mm
Standard
Product No.
Description
Pricing
Lansoprazole
≥98% (TLC), powder
CONDITIONS
column
CYCLOBOND I 2000 DMP, 25 cm x 4.6 mm I.D., 5 μm particles (20724AST)
mobile phase
[A] acetonitrile; [B] methanol; [C] acetic acid; [D] ammonium hydroxide; (95:5:0.4:0.1, A:B:C:D)
flow rate
1 mL/min
column temp.
25 °C
detector
UV, 254 nm
injection
10 μL
sample
1 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
null