MilliporeSigma
HPLC Analysis of Ritalinic Acid and Ritalin (Methylphenidate) Enantiomers on Astec® CHIROBIOTIC® V2 with T2 Guard Column
HPLC Analysis of Ritalinic Acid and Ritalin (Methylphenidate) Enantiomers on Astec® CHIROBIOTIC® V2 with T2 Guard Column application for HPLC
Materials
Analytical column
Product No.
Description
Pricing
Astec® CHIROBIOTIC® V2 Chiral HPLC Column
5 μm particle size, L × I.D. 15 cm × 4.6 mm
Standard
Product No.
Description
Pricing
Methylphenidate hydrochloride
Ritalinic acid
99% (CP)
CONDITIONS
column
CHIROBIOTIC T2 guard column, 2 cm x 4 mm I.D., 5 μm particles (16100AST (requires holder 21150AST))
column
CHIROBIOTIC V2, 15 cm x 4.6 mm I.D., 5 μm particles (15023AST)
mobile phase
[A] 20 mM ammonium acetate, pH 4.1; [B] methanol; (7:93, A:B)
flow rate
1 mL/min
column temp.
25 °C
detector
UV, 230 nm
injection
2 μL
sample
1 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC