MilliporeSigma
HPLC Analysis of Clenbuterol Enantiomers on Astec® CHIROBIOTIC® V
HPLC Analysis of Clenbuterol Enantiomers on Astec® CHIROBIOTIC® V application for HPLC
Materials
Analytical column
Product No.
Description
Pricing
Astec® CHIROBIOTIC® V Chiral HPLC Column
5 μm particle size, L × I.D. 25 cm × 4.6 mm
Application
Product No.
Description
Pricing
LC/MS Analysis of Clenbuterol In Plasma on Astec® CHIROBIOTIC® T with Phospholipid Removal using HybridSPE® Phospholipid
application for SPE, application for LC-MS
LC/MS Analysis of Clenbuterol Enantiomers in Plasma on Astec® CHIROBIOTIC® T after SPE using HybridSPE®-Phospholipid
application for HPLC
HPLC Analysis of Clenbuterol Enantiomers on Astec® CHIROBIOTIC® T
application for HPLC
HPLC Analysis of Clenbuterol Enantiomers on Astec® CHIROBIOTIC® T2
application for HPLC
Standard
Product No.
Description
Pricing
Clenbuterol hydrochloride
≥95%
CONDITIONS
column
CHIROBIOTIC V, 25 cm x 4.6 mm I.D., 5 μm particles (11024AST)
mobile phase
[A] methanol; [B] acetic acid; [C] triethylamine; (99.98:0.01:0.01, A:B:C)
flow rate
1 mL/min
column temp.
25 °C
detector
UV, 254 nm
injection
5 μL
sample
1 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC