MilliporeSigma
HPLC Analysis of Dopa and Carbidopa Enantiomers on Astec® CHIROBIOTIC® T
HPLC Analysis of Dopa and Carbidopa Enantiomers on Astec® CHIROBIOTIC® T application for HPLC
Materials
Analytical column
Product No.
Description
Pricing
Astec® CHIROBIOTIC® T Chiral HPLC Column
5 μm particle size, L × I.D. 25 cm × 4.6 mm
Standard
Product No.
Description
Pricing
S-(−)-Carbidopa
≥98% (TLC), powder
3,4-Dihydroxy-D-phenylalanine
powder, ≥95%
3,4-Dihydroxy-DL-phenylalanine
3,4-Dihydroxy-L-phenylalanine
≥98% (TLC)
CONDITIONS
column
CHIROBIOTIC T, 25 cm x 4.6 mm I.D., 5 μm particles (12024AST)
mobile phase
[A] 20 mM ammonium acetate, pH 4; [B] methanol; (30:70, A:B)
flow rate
1 mL/min
column temp.
25 °C
detector
UV, 230 nm
injection
5 μL
sample
1 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC