MilliporeSigma
HPLC Analysis of Clenbuterol Enantiomers on Astec® CHIROBIOTIC® T
HPLC Analysis of Clenbuterol Enantiomers on Astec® CHIROBIOTIC® T application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Astec® CHIROBIOTIC® T Chiral HPLC Column
5 μm particle size, L × I.D. 25 cm × 4.6 mm
application
Product No.
Description
Pricing
LC/MS Analysis of Clenbuterol In Plasma on Astec® CHIROBIOTIC® T with Phospholipid Removal using HybridSPE® Phospholipid
application for SPE, application for LC-MS
LC/MS Analysis of Clenbuterol Enantiomers in Plasma on Astec® CHIROBIOTIC® T after SPE using HybridSPE®-Phospholipid
application for HPLC
HPLC Analysis of Clenbuterol Enantiomers on Astec® CHIROBIOTIC® T2
application for HPLC
HPLC Analysis of Clenbuterol Enantiomers on Astec® CHIROBIOTIC® V
application for HPLC
standard
Product No.
Description
Pricing
Clenbuterol hydrochloride
≥95%
CONDITIONS
column
CHIROBIOTIC T, 25 cm x 4.6 mm I.D., 5 μm particles (12024AST)
mobile phase
15 mM ammonium formate in methanol
flow rate
1 mL/min
column temp.
25 °C
detector
UV, 220 nm
injection
3 μL
sample
1 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC