MilliporeSigma
HPLC Analysis of Bupivacaine Enantiomers on Astec® CHIROBIOTIC® V2, Formate Mobile Phase
HPLC Analysis of Bupivacaine Enantiomers on Astec® CHIROBIOTIC® V2, Formate Mobile Phase application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Astec® CHIROBIOTIC® V2 Chiral HPLC Column
5 μm particle size, L × I.D. 25 cm × 4.6 mm
application
Product No.
Description
Pricing
HPLC Analysis of Bupivacaine Enantiomers on Astec® CHIROBIOTIC® V2, Acetate Mobile Phase
application for HPLC
HPLC Analysis of Bupivacaine Enantiomers on Astec® CHIROBIOTIC® V
application for HPLC
standard
Product No.
Description
Pricing
Bupivacaine hydrochloride monohydrate
analytical standard, for drug analysis
Bupivacaine-(butyl-1-13C)
99 atom % 13C
CONDITIONS
column
CHIROBIOTIC V2, 25 cm x 4.6 mm I.D., 5 μm particles (15024AST)
mobile phase
[A] 20 mM ammonium formate, pH 4.1; [B] methanol; (5:95, A:B)
flow rate
1 mL/min
column temp.
20 °C
detector
UV, 220 nm
injection
5 μL
sample
1 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC