HPLC Analysis of Fat Soluble Vitamins by Normal Phase Chromatography on Ascentis® Si, 15 cm Column
Materials
Analytical column
Product No.
Description
Pricing
CONDITIONS
column
Ascentis Si, 15 cm × 4.6 mm I.D., 5 μm particles (581512-U)
mobile phase
[A] hexane, [B] ethyl acetate
gradient
10 to 30% B in 10 min; held for 2 min; to 10% B in 1 min
flow rate
1.0 mL/min
column temp.
30 °C
detector
UV, 290 nm
injection
10 μL
sample
as indicated in 96:4 (hexane:isopropanol)
Description
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany