MilliporeSigma
HPLC Analysis of Cis-(+/-)-3,3a,8,8a-Tetrahydro-2H-indeno[1,2-d]oxazol-2-one Enantiomers on Astec® (R,R) P-CAP-DP
HPLC Analysis of Cis-(+/-)-3,3a,8,8a-Tetrahydro-2H-indeno[1,2-d]oxazol-2-one Enantiomers on Astec® (R,R) P-CAP™-DP application for HPLC
Materials
standard
Product No.
Description
Pricing
(3aS-cis)-(−)-3,3a,8,8a-Tetrahydro-2H-indeno[1,2-d]oxazol-2-one
98%
CONDITIONS
column
Astec (R,R) P-CAP-DP, 25 cm x 4.6 mm I.D., 5 μm particles (35024AST)
mobile phase
[A] acetonitrile; [B] methanol; (98:2, A:B)
flow rate
1 mL/min
column temp.
25 °C
detector
UV, 230 nm
injection
1 μL
sample
1 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
P-CAP is a trademark of Sigma-Aldrich Co. LLC