MilliporeSigma
HPLC Analysis of Barbiturates on Discovery® C18
HPLC Analysis of Barbiturates on Discovery® C18 application for HPLC
Materials
Analytical column
Product No.
Description
Pricing
SUPELCOSIL LC-ABZ HPLC Column
5 μm particle size, L × I.D. 15 cm × 4.6 mm
CONDITIONS
column
Discovery C18, 15 cm × 4.6 mm I.D., 5 μm particles (504955)
mobile phase
[A] acetonitrile; [B] 25 mM potassium phosphate buffer, pH 6.9 (25:75, A:B)
flow rate
1.5 mL/min
column temp.
ambient
detector
UV, 254 nm
injection
25 μL
Description
Legal Information
Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany