HPLC Analysis of Aflatoxins on Ascentis® Express C18 with Fluorescence Detection
![HPLC Analysis of Aflatoxins on Ascentis® Express C18 with Fluorescence Detection application for HPLC](/deepweb/assets/sigmaaldrich/product/images/204/284/26ecc5d1-713c-4536-9009-0a8b9da7e82e/800/26ecc5d1-713c-4536-9009-0a8b9da7e82e.jpg)
CONDITIONS
column
Ascentis Express C18, 15 cm x 4.6 mm I.D., 2.7 μm particles (53829-U)
mobile phase
[A] water; [B] acetonitrile; (75:25, A:B)
flow rate
2.0 mL/min
pressure
6381 psi (440 bar)
column temp.
35 °C
detector
fluorescence, ex 360 nm, em 440 nm
injection
50 μL
sample
200 μL aflatoxin standard in acetonitrile; 0.5 μg/mL B2 and G2; 2 μg/mL B1 and G1; 800 μL delivatization solution: 70% water, 20% TFA and 10% acetic acid
heat for 9 min at 65 °C
Description
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany