MilliporeSigma
HPLC Analysis of Polyethylene Glycol 200 on TSKgel® G-OLIGO-PW
HPLC Analysis of Polyethylene Glycol 200 on TSKgel® G-OLIGO-PW application for HPLC
Materials
Analytical column
Product No.
Description
Pricing
TSKgel® G-Oligo-PW HPLC Column
phase hydroxylated methacrylate, L × I.D. 30 cm × 7.8 mm, 7 μm particle size
standard
Product No.
Description
Pricing
Polyethylene Glycol 200
certified reference material, pharmaceutical secondary standard
CONDITIONS
column
TSKgel G-OLIGO-PW, 30 cm × 7.8 mm I.D., 6 μm particles (808031)
mobile phase
distilled water
flow rate
1 mL/min
detector
RI
Description
Legal Information
TSKgel is a registered trademark of Tosoh Corporation