MilliporeSigma
HPLC Analysis of Carboxyamido Methylated BSA Tryptic Digest on Discovery® RP-Amide C16
HPLC Analysis of Carboxyamido Methylated BSA Tryptic Digest on Discovery® RP-Amide C16 application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Discovery® RP-AmideC16 HPLC Column
5 μm particle size, L × I.D. 15 cm × 4.6 mm
CONDITIONS
column
Discovery RP-AmideC16, 25 cm × 4.6 mm I.D., 5 μm particles (505013)
mobile phase
[A] 0.1% TFA, 1% acetonitrile in water; [B] 0.1% TFA, 40% acetonitrile
gradient
0-100% B in 130 min
flow rate
1 mL/min
column temp.
35 °C
detector
UV, 215 nm
Description
Legal Information
Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany