MilliporeSigma
HPLC Analysis of Pesticides on Ascentis® C8 (15 cm x 4.6 mm x 5 μm)
HPLC Analysis of Pesticides on Ascentis® C8 (15 cm x 4.6 mm x 5 μm) application for HPLC
Materials
Analytical column
Product No.
Description
Pricing
Ascentis® C18 HPLC Column
3 μm particle size, L × I.D. 15 cm × 4.6 mm
Ascentis® C18 HPLC Column
5 μm particle size, L × I.D. 25 cm × 4.6 mm
Ascentis® C8 HPLC Column
3 μm particle size, L × I.D. 15 cm × 4.6 mm
Ascentis® C8 HPLC Column
5 μm particle size, L × I.D. 25 cm × 4.6 mm
Ascentis® Express C18, 2.7 μm HPLC Column
2.7 μm particle size, L × I.D. 15 cm × 4.6 mm
Ascentis® Express C8, 2.7 μm HPLC Column
2.7 μm particle size, L × I.D. 15 cm × 4.6 mm
accessory
Product No.
Description
Pricing
Wheaton wide-mouth bottle
amber soda-lime glass bottle, capacity 1000 mL, white polypropylene cap, (PTFE faced LDPE liner), case of 12 ea
Coupler for Legacy Guard Column Holder
PEEK, I.D. × O.D. × Overall L 0.010 in. × 1/16 in. × 1 in.
Supelguard Guard Cartridge Holder
Stand-alone, for use with Supelguard cartridges (2 cm L. x 2.1 to 4.0 mm I.D.)
guard cartridge
Product No.
Description
Pricing
Ascentis® C18 Supelguard Guard Cartridge
5 μm particle size, L × I.D. 2 cm × 4 mm, pkg of 2 ea
Ascentis® C18 Supelguard Guard Cartridge
3 μm particle size, L × I.D. 2 cm × 4 mm, pkg of 2 ea
Ascentis® C8 Supelguard Guard Cartridge
5 μm particle size, L × I.D. 2 cm × 4 mm, pkg of 2 ea
CONDITIONS
column
Ascentis C8, 15 cm x 4.6 mm I.D., 5 μm particles (581424-U)
mobile phase
[A] water; [B] acetonitrile
gradient
35 to 75% B in 13 min; held for 5 min
flow rate
1.0 mL/min
column temp.
35 °C
detector
UV, 254 nm
injection
100 μL
sample
as indicated in 2.2% 90:10 acetonitrile:acetone in DI water
Description
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany