MilliporeSigma
GC Analysis of Glycols on Nukol
GC Analysis of Glycols on Nukol suitable for GC
Materials
analytical column
Product No.
Description
Pricing
Nukol Capillary GC Column
size × I.D. 15 m × 0.53 mm, df 0.50 μm
standard
Product No.
Description
Pricing
Diethylene glycol
Pharmaceutical Secondary Standard; Certified Reference Material
Ethylene glycol
Pharmaceutical Secondary Standard; Certified Reference Material
Propylene glycol
Pharmaceutical Secondary Standard; Certified Reference Material
Glycerin
Pharmaceutical Secondary Standard; Certified Reference Material
2,3-Butanediol
analytical standard, mixture of racemic and meso forms
Triethylene glycol
United States Pharmacopeia (USP) Reference Standard
CONDITIONS
column
Nukol, 15 m x 0.53 mm I.D., 0.5 μm (25326)
oven
110 °C, 8 °C/min to 220 °C
detector
FID, 250 °C
carrier gas
helium, 15 mL/min
injection
1 μL, direct
sample
100 ppm each glycol in water
Description
Legal Information
null