MilliporeSigma
GC Analysis of Volatiles in Milk (Before and After Exposure to Sunlight) on Supel-Q PLOT after SPME using a 75 μm PDMS/Carboxen Fiber
GC Analysis of Volatiles in Milk (Before and After Exposure to Sunlight) on Supel-Q PLOT after SPME using a 75 μm PDMS/Carboxen Fiber suitable for GC, application for SPME
Materials
analytical column
Product No.
Description
Pricing
Supel-Q PLOT Capillary GC Column
L × I.D. 30 m × 0.32 mm, average thickness 15 μm
SPME fiber
Product No.
Description
Pricing
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS)
df 75 μm(CAR/PDMS, for use with manual holder, needle size 24 ga
related product
Product No.
Description
Pricing
SPME Fiber Holder
for use with manual sampling
SPME Fiber Holder
for use with CTC CombiPAL and Gerstel MPS2
standard
Product No.
Description
Pricing
4-Methyl-2-pentanone
analytical standard
Hexanal
analytical standard
Dimethyl disulfide
analytical standard
CONDITIONS
sample/matrix
3 g of 2% milk + 10 μL internal standard solution (20 μg/mL 4-methyl-2-pentanone) (9 mL GC vial)
SPME fiber
PDMS/Carboxen, 75 μm (57318)
extraction
headspace, 15 min with constant stirring at 45 °C
desorption process
5 min, 250 °C
column
Supel-Q PLOT, 30 m × 0.32 mm I.D. (24242)
oven
70 °C (2 min) to 140 °C at 6 °C/min (2 min hold) then to 220 °C at 6 °C/min (5 min hold)
inj. temp.
250 °C
detector
GC-MS ion trap, m/z = 33-300
carrier gas
helium, 35 cm/sec
injection
splitless, closed 2 min
liner
0.75 mm liner
sample
Milk
Description
Analysis Note
In this study, SPME was used for the analysis of volatiles in milk using a headspace method. Light can catalyze the formation of peroxides that can cleave double bonds in fatty acids. The cleaved by-products form aldehydes that can give the food a pungent off flavor. Dr. Ray Marsili investigated the feasibility of using SPME as a substitute sample preparation device prior to GC/MS analysis. The Carboxen/PDMS fiber was evaluated for the extraction of low ppb levels of pentanal, hexanal, and dimethyldisulfide. The results of the comparison of SPME to dynamic headspace analysis indicate that SPME-GC/MS is more accurate and precise while providing sensitivities equivalent to DH-GC/MS. The GC column used, Supel-Q PLOT, operates via gas-solid chromatography with a DVB adsorbent and permits the separation of a wide range of analytes.
Legal Information
null