MilliporeSigma
GC Analysis of δ-Decanolactone Enantiomers on Astec® CHIRALDEX G-TA
GC Analysis of δ-Decanolactone Enantiomers on Astec® CHIRALDEX™ G-TA suitable for GC
Materials
Analytical column
Product No.
Description
Pricing
Astec® CHIRALDEX G-TA Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.12 μm
standard
Product No.
Description
Pricing
δ-Decalactone
≥98%, FCC, FG
δ-Decalactone
natural, ≥98%, FCC, FG
CONDITIONS
column
Astec CHIRALDEX G-TA, 30 m x 0.25 mm I.D., 0.12 μm (73033AST)
oven
helium, 24 psi
inj. temp.
250 °C
detector
FID, 250 °C
carrier gas
helium, 24 psi
sample
peaks 1 & 2: δ-decanolactone enantiomers
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIRALDEX is a trademark of Sigma-Aldrich Co. LLC