GC Analysis of Potpourri Fragrance Compounds on SLB®-5ms
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
SLB-5ms, 30 m x 0.25 mm I.D., 0.25 μm (28471-U)
oven
50 °C (0 min), then 8 °C/min to 200 °C (1 min) then 20 °C/min to 300 °C (20 min)
inj. temp.
280 °C
detector
FID, 310 °C
carrier gas
helium, 25cm/ sec set at 110°C
injection
0.1 μL 100:1 split
liner
4 mm I.D. cup injector
sample
Mixture of Potpourri Fragrance Compounds, neat
Description
Legal Information
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany