MilliporeSigma
GC Analysis of 2-Halohydrocarbon Enantiomers on Astec® CHIRALDEX G-TA
GC Analysis of 2-Halohydrocarbon Enantiomers on Astec® CHIRALDEX™ G-TA suitable for GC
Materials
analytical column
Product No.
Description
Pricing
Astec® CHIRALDEX G-TA Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.12 μm
Standard
Product No.
Description
Pricing
2-Bromopentane
95%
2-Chlorobutane
≥99%
2-Bromobutane
98%
CONDITIONS
column
Astec CHIRALDEX G-TA, 30 m x 0.25 mm I.D., 0.12 μm (73033AST)
gradient
30 °C (3 min), 5 °C/min to 70 °C
inj. temp.
250 °C
detector
FID, 250 °C
carrier gas
helium, 30 psi
injection
1 μL, 80:1 split
sample
2.5 mg/mL of each compound
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIRALDEX is a trademark of Sigma-Aldrich Co. LLC