MilliporeSigma
US EPA Method TO-10/IP-8 and ASTM® D4861: GC Analysis of Pesticides on SPB®-608 after Collection/Desorption using ORBO-1000
US EPA Method TO-10/IP-8 and ASTM® D4861: GC Analysis of Pesticides on SPB®-608 after Collection/Desorption using ORBO™-1000 suitable for GC, application for air monitoring
Materials
Analytical column
Product No.
Description
Pricing
SPB®-608 Capillary GC Column
L × I.D. 30 m × 0.53 mm, df 0.50
Related product
Product No.
Description
Pricing
ORBO 1000 Precleaned Small PUF Cartridge
O.D. × L 22 mm × 7.6 cm, pkg of 3 ea
CONDITIONS
sample/matrix
polyurethane foam (PUF) spiked with 250 ng each pesticide
adsorbent tube
ORBO-1000 (20557)
column
SPB-608, 30 m × 0.53 mm I.D., 0.5 μm film (25312)
oven
150 °C (1 min) to 270 °C at 4 °C/min
detector
ECD
carrier gas
helium, 7 mL/min
injection
1 μL
Description
Legal Information
ASTM is a registered trademark of American Society for Testing and Materials
ORBO is a trademark of Sigma-Aldrich Co. LLC
SPB is a registered trademark of Merck KGaA, Darmstadt, Germany