MilliporeSigma
US EPA Method 8260: GC Analysis of Volatiles on VOCOL® after Purge & Trap using "K" Trap
US EPA Method 8260: GC Analysis of Volatiles on VOCOL® after Purge & Trap using "K" Trap suitable for GC
Materials
Analytical column
Product No.
Description
Pricing
VOCOL® Capillary GC Column
L × I.D. 60 m × 0.25 mm, df 1.50 μm
VOCOL® Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 1.50 μm
SPB®-624 Capillary GC Column
L × I.D. 60 m × 0.25 mm, df 1.40 μm
SPB®-624 Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 1.40 μm
VOCOL® Capillary GC Column
L × I.D. 20 m × 0.18 mm, df 1.00 μm
SPB®-624 Capillary GC Column
L × I.D. 20 m × 0.18 mm, df 1.00 μm
accessory
Product No.
Description
Pricing
Hamilton® syringe, 1000 series GASTIGHT®, PTFE luer lock
1005TLL, PTFE Luer lock, volume 5 mL, needle size (not included)
Soil Sampler
size 150 mm × 19 mm, Tekmar® 12-0507-024
Fritless Purge Sampler Glassware
size 5 mL, Tekmar® 14-2336-024
Fritted Purge Sampler
size 5 mL, Tekmar® 14-2337-024
Fritted Purge Sampler
size 5 mL,
right stem
, Tekmar® 14-3544-124
Hamilton® syringe, 1000 series GASTIGHT®, PTFE luer lock
1005SLPT (for Purge & Trap), volume 5 mL, needle size (not included)
related product
Product No.
Description
Pricing
EPA Vials, white polypropylene screw top hole cap, pre-cleaned and assembled
volume 40 mL, clear glass vial, vial O.D. × H 29 mm × 82 mm, pkg of 72 ea
EPA Vials, white polypropylene screw top hole cap, pre-cleaned and assembled
volume 40 mL, amber glass vial, O.D. × H 29 mm × 82 mm, pkg of 72 ea
sample prep
Product No.
Description
Pricing
Purge/Trap K Vocarb® 3000
for use with Tekmar® Velocity XPT, LSC-1, LSC-2, LSC-2000, 4000 (Straight, 12 in./30.5 cm, Swagelok fitting), HT-3
Purge/Trap K Vocarb® 3000
for use with Dynatech "Dyna"
Purge/Trap K Vocarb® 3000
for use with OI Analytical 4460
Vocarb® 3000 Purge Trap K, CDS
for use with CDS Peakmaster
Purge/Trap K Vocarb® 3000
for use with Tekmar® 3000, 3100 (Straight, 12 in./30.5 cm, Valco fitting
Purge/Trap K Vocarb® 3000
for use with OI Analytical Eclipse 4660, 4560
standard
Product No.
Description
Pricing
Perfluorotributylamine (PFTBA)
analytical standard
EPA 8260A Surrogate Standards Mix
certified reference material, 2000 μg/mL each component in methanol
EPA 8240B/8260A Matrix Spike Mix
certified reference material, 2500 μg/mL each component in methanol
4-Bromofluorobenzene solution
certified reference material, 2000 μg/mL in methanol
EPA 8260 Volatiles Calibration Mix
certified reference material, 2000 μg/mL each component in methanol
EPA 8260 Internal Standards Mix 1
certified reference material, 2500 μg/mL in methanol
Methyl tert-butyl ether solution
certified reference material, TraceCERT®
TCL Volatiles Mix
certified reference material, 2000 μg/mL each component in methanol: water (9:1)
CONDITIONS
sample/matrix
each analyte at 50 ppb in 5 mL water
purge trap
VOCARB 3000 “K”
purge
40 mL/min at 25 °C for 11 min
dry purge
2 min
desorption preheat
205 °C
desorption process
40 mL/min at 210 °C for 2 min
bake
260 °C for 10 min
transfer line/valve temp.
110 °C
column
VOCOL, 30 m × 0.25 mm I.D., 1.5 μm (24205-U)
oven
40 °C (2 min), 7 °C/min to 125 °C, 12 °C/min to 220 °C (5 min)
inj. temp.
150 °C
MSD interface
200 °C
scan range
m/z = 35-400
carrier gas
helium, 0.7 mL/min
injection
30:1 split
liner
0.75 mm I.D. SPME
Description
Legal Information
VOCOL is a registered trademark of Merck KGaA, Darmstadt, Germany