MilliporeSigma
US EPA Method 525.2: GC Analysis of Semivolatiles on SLB®-5ms
US EPA Method 525.2: GC Analysis of Semivolatiles on SLB®-5ms suitable for GC
Materials
Analytical column
Product No.
Description
Pricing
SLB®-5ms Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.25 μm
CONDITIONS
column
SLB-5ms, 30 m x 0.25 mm I.D., 0.25 μm (28471-U)
oven
45 °C (1 min), 50 °C/min to 130 °C (1 min), 12 °C/min to 180 °C, 7 °C/min to 240 °C, 12 °C/min to 330 °C (2 min)
inj. temp.
275 °C
MSD interface
330 °C
scan range
m/z = 45-450
carrier gas
helium, 0.9 mL/min., constant flow
injection
1 μL pulsed splitless (15 psi until 0.20 min) (1.0 min)
liner
4 mm I.D., single taper
sample
2 ppm, in ethyl acetate, of a 102 component semivolatile standard plus 3 surrogate and 3 internal standard compounds (at 5 ppm)
Description
Legal Information
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany