MilliporeSigma
GC Analysis of Total Petroleum Hydrocarbons (TPH) on SLB®-5ms (20 m x 0.18 mm I.D., 0.18 μm), Fast GC Analysis
GC Analysis of Total Petroleum Hydrocarbons (TPH) on SLB®-5ms (20 m x 0.18 mm I.D., 0.18 μm), Fast GC Analysis suitable for GC
Materials
analytical column
Product No.
Description
Pricing
SLB®-5ms Capillary GC Column
L × I.D. 20 m × 0.18 mm, df 0.18 μm
CONDITIONS
column
SLB-5ms, 20 m x 0.18 mm I.D., 0.18 μm (28564-U)
oven
70 °C (0.5 min), 20 °C/min to 360 °C (1 min)
inj. temp.
275 °C
detector
FID, 365 °C
carrier gas
hydrogen, 2.5 mL/min constant
injection
0.50 μL, 200:1 split
liner
2 mm I.D. straight
sample
TPH Mix 3, each analyte at 1000 μg/mL in carbon disulfide (861394-U)
Description
Legal Information
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany