MilliporeSigma
GC Analysis of Industrial Solvents on SUPELCOWAX 10
GC Analysis of Industrial Solvents on SUPELCOWAX 10 suitable for GC
Materials
Analytical column
Product No.
Description
Pricing
SUPELCOWAX 10 Capillary GC Column
L × I.D. 30 m × 0.32 mm, df 1.00 μm
CONDITIONS
column
SUPELCOWAX 10, 30 m x 0.32 mm I.D., 1.0 μm (24211)
oven
35 °C (8 min), 4 °C/min to 130 °C (2 min)
inj. temp.
250 °C
detector
FID, 250 °C
carrier gas
helium, 25 cm/sec
injection
0.5 μL, 200:1 split
liner
split, cup design
sample
59 component neat solvent mixture
Description
Legal Information
null