MilliporeSigma
GC Analysis of Hydrocarbons in Gasoline on Petrocol® DH, Isothermal
GC Analysis of Hydrocarbons in Gasoline on Petrocol® DH, Isothermal suitable for GC
Materials
Analytical column
Product No.
Description
Pricing
Petrocol® DH Capillary GC Column
L × I.D. 100 m × 0.25 mm, df 0.50 μm
CONDITIONS
column
Petrocol DH, 100 m x 0.25 mm I.D., 0.50 μm (24160-U)
oven
60 °C
inj. temp.
250 °C
detector
FID, 250 ºC
carrier gas
helium, 19-21 cm/sec
injection
0.1 μL, 100:1 split
Description
Analysis Note
Detailed hydrocarbon analysis (DHA) is performed on long non-polar columns. Elution occurs in order of lowest boiling point to highest. The chromatogram shown here is of a gasoline sample using a 100 m Petrocol DH column. The long column length and 60 ºC isothermal oven temperature allow the separation of many components in this complex sample. Compounds identified range from propane (peak 1) to 1-methylnaphthalene (peak 96). Retention index libraries exist for this column, adding to its usefulness.
Legal Information
Petrocol is a registered trademark of Merck KGaA, Darmstadt, Germany