MilliporeSigma
GC Analysis of a 37-Component FAME Mix on SP®-2560 (75 m x 0.18 mm I.D., 0.14 μm), Fast GC Analysis
GC Analysis of a 37-Component FAME Mix on SP®-2560 (75 m x 0.18 mm I.D., 0.14 μm), Fast GC Analysis suitable for GC
Materials
analytical column
Product No.
Description
Pricing
SP®-2560 Capillary GC Column
L × I.D. 75 m × 0.18 mm, df 0.14 μm
CONDITIONS
column
SP-2560, 75 m x 0.18 mm I.D., 0.14 μm (23348-U)
oven
140 °C (5 min), 4 °C/min to 240 °C (2 min)
inj. temp.
250 °C
detector
FID, 250 °C
carrier gas
hydrogen, 40 cm/sec. @ 175 °C
injection
1 μL, split 100:1
liner
4 mm I.D split, cup design
sample
37 component FAME mix at concentrations listed in methylene chloride (47885-U)
Description
Legal Information
SP is a registered trademark of Sigma-Aldrich Co. LLC