MilliporeSigma
EN Method 2005/108/EC: GC Analysis of Pesticide/PAH Compounds on SLB®-5ms
EN Method 2005/108/EC: GC Analysis of Pesticide/PAH Compounds on SLB®-5ms suitable for GC
Materials
analytical column
Product No.
Description
Pricing
SLB®-5ms Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.25 μm
CONDITIONS
column
SLB-5ms, 30 m x 0.25 mm I.D., 0.25 μm (28471-U)
oven
50 °C (3 min), 25 °C/min to 250 °C (3 min), 4 °C/min to 325 °C (5 min)
inj. temp.
300 °C
MSD interface
300 °C / 200 °C (source/quads)
scan range
SIM
carrier gas
helium, 0.9 mL/min constant
injection
0.5 μL, pulsed (15 psi until 0.20 min) splitless (1.0 min)
liner
2 mm I.D. FocusLiner inlet liner with taper
sample
26 analytes at 50 ppb each in methylene chloride
Description
Legal Information
FocusLiner is a trademark of SGE Analytical Science Pty Ltd.
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany