MilliporeSigma
  • Presence of NGF and its receptor TrkA in degenerative lumbar facet joint specimens.
Presence of NGF and its receptor TrkA in degenerative lumbar facet joint specimens.
European spine journal : official publication of the European Spine Society, the European Spinal Deformity Society, and the European Section of the Cervical Spine Research Society (2009-04-29)
M F Surace, D Prestamburgo, M Campagnolo, A Fagetti, L Murena
ABSTRACT
In a preliminary study, the recurrent presence of nervous terminations was demonstrated with optical microscopy in several slides of degenerative lumbar facet joints and surrounding soft tissues. The purpose of this study was to prove the presence of NGF (nerve growth factor) and its receptor TrkA (tyrosine kinase receptor) with immunofluorescence. The peri/articular tissues were harvested from the lumbar facet joints of ten patients surgically treated for degenerative diseases. There were seven females (one bilateral) and two males whose mean age at surgery was 72 years (range, 67-80 years). The affected levels were L3-L4 in two cases and L4-L5 in seven cases (one bilateral). All specimens were fixed in formalin, dehydrated and enclosed in paraffin. From each specimen, four slides were obtained. Two slides were employed for the search of NGF: one was treated with specific antibodies and marked with FITC (fluorescein isothiocyanate conjugated), and the second slide was for control purposes. It was exposed to FITC, but without prior exposure to the specific antibody. The same procedure was repeated to obtain on two more slides, to repeat the search for Trka with specific antibodies. All the slides were finally studied on a fluoromicroscope. The analysis of these specimens revealed the presence of the neurotrophin (NGF) and its own receptor (TrkA) in all cases: the immunohistochemical reaction between the specimens and the specific antibodies marked with FITC was seen under fluoromicroscopy, but in none of the control cases treated with FITC only. NGF is released by mastocytes, fibroblasts and other cell types involved in the inflammatory processes. The level of peripheral NGF is increased in inflammatory processes, while the administration of exogenous NGF has a hyperalgesic effect on rats and produces muscular pain in humans. Furthermore, NGF produces hypersensitization to heat stimulation in humans and mammals in general. There is considerable evidence showing that the system constituted by the NGF and its high-affinity receptor TrkA plays a fundamental role in the molecular processes underlying the main forms of "persistent" pain. This indicates a possible therapeutic area for the antibodies that could block the NGF/TrkA system, in order to modulate the frequency and the duration of the action potential of nociceptive neurons during chronic inflammation. This study demonstrated the presence of NGF and TrkA in specimens collected from degenerative facet joints, suggesting that specific molecules could be used in order to modulate chronic pain in patients with degenerative lumbar spine.