MilliporeSigma
  • K-ras mutations in lung tumors and tumors from other organs are consistent with a common mechanism of ethylene oxide tumorigenesis in the B6C3F1 mouse.
K-ras mutations in lung tumors and tumors from other organs are consistent with a common mechanism of ethylene oxide tumorigenesis in the B6C3F1 mouse.
Toxicologic pathology (2007-02-28)
Hue-Hua L Hong, Christopher D Houle, Thai-Vu T Ton, Robert C Sills
ABSTRACT
Ethylene oxide is a multisite carcinogen in rodents and classified as a human carcinogen by the National Toxicology Program. In 2-year mouse studies, ethylene oxide (EO) induced lung, Harderian gland (HG), and uterine neoplasms. We evaluated representative EO-induced and equivalent spontaneous neoplasms for K-ras mutations in codons 12, 13, and 61. K-ras mutations were identified in 100% (23/23) of the EO-induced lung neoplasms and 25% (27/108) of the spontaneous lung neoplasms. Codon 12 G to T transversions were common in EO-induced lung neoplasms (21/23) but infrequent in spontaneous lung neoplasms (1/108). K-ras mutations were found in 86% (18/21) of the EO-induced HG neoplasms and 7% (2/27) of the spontaneous HG neoplasms. Codon 13 G to C and codon 12 G to T transversions were predominant in the EO-induced HG neoplasms but absent in spontaneous HG neoplasms (0/27). K-ras mutations occurred in 83% (5/6) of the EO-induced uterine carcinomas and all were codon 13 C to T transitions. These data show a strong predilection for development of K-ras mutations in EO-induced lung, Harderian gland, and uterine neoplasms. This suggests that EO specifically targets the K-ras gene in multiple tissue types and that this event is a critical component of EO-induced tumorigenesis.