MilliporeSigma
  • MicroRNA-92a-3p Regulates Aggrecanase-1 and Aggrecanase-2 Expression in Chondrogenesis and IL-1β-Induced Catabolism in Human Articular Chondrocytes.
MicroRNA-92a-3p Regulates Aggrecanase-1 and Aggrecanase-2 Expression in Chondrogenesis and IL-1β-Induced Catabolism in Human Articular Chondrocytes.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology (2017-12-15)
Guping Mao, Peihui Wu, Ziji Zhang, Zhiqi Zhang, Weiming Liao, Yukang Li, Yan Kang
ABSTRACT
Aggrecanase-1 (ADAMTS-4) and aggrecanase-2 (ADAMTS-5) are secreted enzymes belonging to the ADAMTS (a disintegrin and metalloproteinase with thrombospondin motifs) family that play significant roles in the progression of osteoarthritis (OA). Here, we aimed to determine whether the expression of ADAMTS-4/5 in chondrogenesis and inflammation is regulated by microRNA-92a-3p (miR-92a-3p). MiR-92a-3p and ADAMTS-4/5 expressions were determined by quantitative polymerase chain reaction (qPCR). To investigate the repressive effect of miR-92a-3p on ADAMTS-4/5 expression, chondrogenic human mesenchymal stem cells (hMSCs) and human chondrocytes were transfected with mature miR-92a-3p or an antisense inhibitor (anti-miR-92a-3p), respectively. ADAMTS-4/5 protein production was quantified by enzyme-linked immunosorbent assay (ELISA), and miR-92a-3p involvement in IL-1β-mediated catabolic effects was examined by immunoblotting. The roles of activated MAP kinases (MAPK) and nuclear factor (NF)-κB were evaluated by using specific inhibitors. Interaction between miR-92a-3p and its putative binding site in the 3'-untranslated region (3'-UTR) of ADAMTS-4/5 mRNA was confirmed by luciferase reporter assay. miR-92a-3p expression was elevated in chondrogenic hMSCs, with significantly lower expression in OA cartilage than in normal cartilage. Stimulation with IL-1β significantly reduced miR-92a-3p expression in primary human chondrocytes (PHCs). Transfection of chondrocytes with miR-92a-3p downregulated IL-1β-induced ADAMTS-4/5 expression, and the activity of a reporter construct containing the 3'-UTR of human ADAMTS-4/5 mRNA. MiR-92a-3p expression was suppressed upon IL-1β-induced activation of MAPK and NF-κB in chondrocytes. MiR-92a-3p is an important regulator of ADAMTS-4/5 in human chondrocytes and may contribute to the development of OA.
Sigma-Aldrich
Human ADAMTS4 ELISA Kit, for cell culture supernatants, plasma, and serum samples