MilliporeSigma
  • Natural and anthropogenic particulate-bound aliphatic and polycyclic aromatic hydrocarbons in surface waters of the Gulf of Gabès (Tunisia, southern Mediterranean Sea).
Natural and anthropogenic particulate-bound aliphatic and polycyclic aromatic hydrocarbons in surface waters of the Gulf of Gabès (Tunisia, southern Mediterranean Sea).
Environmental science and pollution research international (2017-11-12)
Rania Fourati, Marc Tedetti, Catherine Guigue, Madeleine Goutx, Hatem Zaghden, Sami Sayadi, Boubaker Elleuch
ABSTRACT
Particulate-bound aliphatic and polycyclic aromatic hydrocarbons (AHs and PAHs) were investigated in the surface waters of the Gulf of Gabès (Tunisia, southern Mediterranean Sea). Samples were collected off the Sfax and Gabès-Ghannouch coasts. Concentrations in total resolved n-alkanes ranged from 0.03 to 3.2 μg L
Product Description
Supelco
Alkane standard solution C21-C40, analytical standard
Pricing and availability is not currently available.