MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Vitamin C exerts novel protective effects against cadmium toxicity in mouse spermatozoa by inducing the dephosphorylation of dihydrolipoamide dehydrogenase.
Vitamin C exerts novel protective effects against cadmium toxicity in mouse spermatozoa by inducing the dephosphorylation of dihydrolipoamide dehydrogenase.
Reproductive toxicology (Elmsford, N.Y.) (2017-11-22)
Lirui Wang, Peifei Li, Yi Wen, Qiangzhen Yang, Linqing Zhen, Jieli Fu, Yuhua Li, Sisi Li, Chengxiao Han, Xinhong Li
ABSTRACT
Cadmium (Cd) has been reported to inhibit mouse sperm motility by inducing the tyrosine phosphorylation of dihydrolipoamide dehydrogenase (DLD). This study aimed to assess the potential effects of vitamin C (Vc) on ameliorating Cd-induced tyrosine phosphorylation of DLD and the specific underlying mechanism. Vc induced the dephosphorylation of DLD or inhibited the tyrosine phosphorylation of DLD. Accordingly, DLD activity, nicotinamide adenine dinucleotide hydrogen (NADH) levels, ATP levels and motility parameters were all restored to normal levels by Vc. Moreover, the effects of Vc on ameliorating these indicators had striking similarities to the effects of ethylenediaminetetraacetic acid (EDTA). In addition, neither the antioxidant melatonin nor the universal oxidant H
Sigma-Aldrich
Adenosine 5′-triphosphate (ATP) bioluminescent somatic cell assay kit, for cellular ATP determination