MilliporeSigma
  • Bcl-3 induced by IL-22 via STAT3 activation acts as a potentiator of psoriasis-related gene expression in epidermal keratinocytes.
Bcl-3 induced by IL-22 via STAT3 activation acts as a potentiator of psoriasis-related gene expression in epidermal keratinocytes.
European journal of immunology (2017-09-14)
Mikiko Tohyama, Yuji Shirakata, Yasushi Hanakawa, Xiuju Dai, Ken Shiraishi, Masamoto Murakami, Saori Miyawaki, Hideki Mori, Ryo Utsunomiya, Kana Masuda, Koji Hashimoto, Koji Sayama
ABSTRACT
IL-22 induces STAT3 phosphorylation and mediates psoriasis-related gene expression. However, the signaling mechanism leading from pSTAT3 to the expression of these genes remains unclear. We focused on Bcl-3, which is induced by STAT3 activation and mediates gene expression. In cultured human epidermal keratinocytes, IL-22 increased Bcl-3, which was translocated to the nucleus with p50 via STAT3 activation. The increases in CXCL8, S100As and human β-defensin 2 mRNA expression caused by IL-22 were abolished by siRNA against Bcl-3. Although CCL20 expression was also augmented by IL-22, the knockdown of Bcl-3 increased its level. Moreover, the combination of IL-22 and IL-17A enhanced Bcl-3 production, IL-22-induced gene expression, and the expression of other psoriasis-related genes, including those encoding IL-17C, IL-19, and IL-36γ. The expression of these genes (except for CCL20) was also suppressed by the knockdown of Bcl-3. Bcl-3 overexpression induced CXCL8 and HBD2 expression but not S100As expression. We also compared Bcl-3 expression between psoriatic skin lesions and normal skin. Immunostaining revealed strong signals for Bcl-3 and p50 in the nucleus of epidermal keratinocytes from psoriatic skin. The IL-22-STAT3-Bcl-3 pathway may be important in the pathogenesis of psoriasis.
Sigma-Aldrich
MISSION® esiRNA, targeting human BCL3