MilliporeSigma
  • Structural Insights into the pH-Dependent Conformational Change and Collagen Recognition of the Human Mannose Receptor.
Structural Insights into the pH-Dependent Conformational Change and Collagen Recognition of the Human Mannose Receptor.
Structure (London, England : 1993) (2017-12-12)
Zhenzheng Hu, Xiangyi Shi, Bowen Yu, Na Li, Ying Huang, Yongning He
ABSTRACT
Mannose receptor (MR, CD206) is an endocytic receptor on microphages and dendritic cells. It recognizes multiple ligands and plays important roles in regulating immune responses and maintaining glycoprotein homeostasis. However, the structure and functional mechanism of MR remain unclear. Here we determine the crystal structures of the N-terminal fragments of MR and reveal the potential binding mode of collagen on the fibronectin II domain. The SAXS and other biophysical data suggest that MR adopts an extended conformation at physiological pH and undergoes conformational changes as pH decreases, resulting in a compact conformation in an acidic environment. Moreover, biochemical data show that MR binds to collagen in a Ca
Sigma-Aldrich
D-(+)-Fucose, ≥97% (GC)