MilliporeSigma
  • Dual functional dinuclear platinum complex with selective reactivity towards c-myc G-quadruplex.
Dual functional dinuclear platinum complex with selective reactivity towards c-myc G-quadruplex.
Scientific reports (2018-01-18)
Lei He, Zhenyu Meng, Dechen Xu, Fangwei Shao
ABSTRACT
G-quadruplexes (GQ) folded by the oncogenic G-rich sequences are the promising targets for developing anticancer therapeutic molecules. However, the current drug development mainly focused on non-covalent dynamic binders to stabilize GQ structures, while the covalent targeting from inorganic complexes via chelating principles, as a potent therapeutic strategy was surprisingly lack of exploration. Herein, a series of dinuclear platinum complexes, [(Pt(Dip)Cl)
Sigma-Aldrich
Thiazole Orange, Dye content ~90 %
Pricing and availability is not currently available.
Sigma-Aldrich
1,10-Diaminodecane, 97%
Pricing and availability is not currently available.