MilliporeSigma
  • Molecular Marker Study of Particulate Organic Matter in Southern Ontario Air.
Molecular Marker Study of Particulate Organic Matter in Southern Ontario Air.
Journal of analytical methods in chemistry (2017-10-28)
Satoshi Irei, Jacek Stupak, Xueping Gong, Tak-Wai Chan, Michelle Cox, Robert McLaren, Jochen Rudolph
ABSTRACT
To study the origins of airborne particulate organic matter in southern Ontario, molecular marker concentrations were studied at Hamilton, Simcoe, and York Gateway Tunnel, representing industrial, rural, and heavy traffic sites, respectively. Airborne particulate matter smaller than 10
Product Description
Supelco
17β(H),21β(H)-Hopane solution, 0.1 mg/mL in isooctane, analytical standard