MilliporeSigma
Broadband
Journal of magnetic resonance (San Diego, Calif. : 1997) (2017-12-16)
Alexander A Marchione, Elizabeth L Diaz
ABSTRACT
A train of BURBOP universal rotation pulses has been used to generate a spin lock sufficient to observe TOCSY correlations over a 46 kHz
Sigma-Aldrich
Perfluorohexyl iodide, 99%
Pricing and availability is not currently available.