- Stonin 2 Overexpression is Correlated with Unfavorable Prognosis and Tumor Invasion in Epithelial Ovarian Cancer.
Stonin 2 Overexpression is Correlated with Unfavorable Prognosis and Tumor Invasion in Epithelial Ovarian Cancer.
International journal of molecular sciences (2017-08-02)
Xiaoying Sun, Weijing Zhang, Han Li, Chunhao Niu, Yulan Ou, Libing Song, Yanna Zhang
PMID28758939
ABSTRACT
Stonin 2 (
Sigma-Aldrich
Anti-STON2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution