MilliporeSigma
  • The dietary flavonoids myricetin and fisetin act as dual inhibitors of DNA topoisomerases I and II in cells.
The dietary flavonoids myricetin and fisetin act as dual inhibitors of DNA topoisomerases I and II in cells.
Mutation research (2009-12-23)
Miguel López-Lázaro, Elaine Willmore, Caroline A Austin
ABSTRACT
DNA topoisomerases (topos) are the target of several drugs commonly used in cancer chemotherapy; these drugs induce topo-DNA complexes with either topo I or topo II that eventually trigger cell death. The inhibition of these enzymes induces DNA alterations that may also lead to carcinogenic effects; indeed, an increased risk for developing leukemia has been observed in patients treated with some topo II inhibitors. Several flavonoids have been shown to interact with purified topo I and topo II, therefore suggesting that these compounds may possess both anticancer and carcinogenic activity. Because the activity of a drug on purified topoisomerases does not always represent the activity in the cell, the aim of this work is to evaluate the effects of several common dietary flavonoids on these enzymes in cells. Using the cell-based TARDIS assay, we have evaluated the effects of the flavonoids quercetin, apigenin, fisetin and myricetin on topo I and topo II in K562 human leukemia cells at several concentrations and exposure times. Quercetin and apigenin induced moderate levels of topo II-DNA complexes and did not induce topo I-DNA complexes in these cells. Fisetin induced neither topo I- nor topo II-DNA complexes, but behaved as a catalytic inhibitor of both enzymes. Myricetin induced high levels of topo-DNA complexes with both enzymes. In addition, murine embryo fibroblasts lacking topo IIbeta were resistant to myricetin-induced cell-growth inhibition, therefore suggesting that topo IIbeta is an important drug target for this flavonoid. These results support the idea that specific concentrations of some dietary flavonoids may produce topoisomerase-mediated carcinogenic and chemotherapeutic effects in vivo. The ability of myricetin to induce topo-DNA complexes with both topo I and topo II in leukemia cells may be therapeutically useful and deserves further study.
Sigma-Aldrich
Myricetin, ≥96.0% (HPLC)