MilliporeSigma
  • Tumor necrosis factor α accelerates Hep-2 cells proliferation by suppressing TRPP2 expression.
Tumor necrosis factor α accelerates Hep-2 cells proliferation by suppressing TRPP2 expression.
Science China. Life sciences (2017-07-02)
Jing Wu, Jizheng Guo, Yunyun Yang, Feifei Jiang, Shuo Chen, Kaile Wu, Bing Shen, Yehai Liu, Juan Du
ABSTRACT
TRPP2, a Ca
Sigma-Aldrich
MISSION® esiRNA, targeting human PKD2
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide, ≥97% (HPLC), powder