MilliporeSigma
Controlling Ag-doping in [Ag
Nanoscale (2017-12-01)
Renxi Jin, Shuo Zhao, Chong Liu, Meng Zhou, Gihan Panapitiya, Yan Xing, Nathaniel L Rosi, James P Lewis, Rongchao Jin
ABSTRACT
Doping metal nanoclusters with a second type of metal is a powerful method for tuning the physicochemical properties of nanoclusters at the atomic level and it also provides opportunities for a fundamental understanding of alloying rules as well as new applications. Herein, we have devised a new, one-phase strategy for achieving heavy Ag-doping in Au
Sigma-Aldrich
2-Methyltetrahydrofuran, BioRenewable, anhydrous, ≥99%, Inhibitor-free