MilliporeSigma
  • Increased circulating interleukin-8 in patients with resistance to thyroid hormone receptor α.
Increased circulating interleukin-8 in patients with resistance to thyroid hormone receptor α.
Endocrine connections (2017-11-05)
Anne H van der Spek, Olga V Surovtseva, Saskia Aan, Anton T J Tool, Annemarie van de Geer, Korcan Demir, Anja L M van Gucht, A S Paul van Trotsenburg, Timo K van den Berg, Eric Fliers, Anita Boelen
ABSTRACT
Innate immune cells have recently been identified as novel thyroid hormone (TH) target cells in which intracellular TH levels appear to play an important functional role. The possible involvement of TH receptor alpha (TRα), which is the predominant TR in these cells, has not been studied to date. Studies in TRα
Sigma-Aldrich
Ampliflu Red, for fluorescence, ≥98.0% (HPLC)
Pricing and availability is not currently available.