MilliporeSigma
Improved detection of synthetic lethal interactions in
Proceedings of the National Academy of Sciences of the United States of America (2017-12-01)
Benjamin E Housden, Zhongchi Li, Colleen Kelley, Yuanli Wang, Yanhui Hu, Alexander J Valvezan, Brendan D Manning, Norbert Perrimon
ABSTRACT
Synthetic sick or synthetic lethal (SS/L) screens are a powerful way to identify candidate drug targets to specifically kill tumor cells, but this approach generally suffers from low consistency between screens. We found that many SS/L interactions involve essential genes and are therefore detectable within a limited range of knockdown efficiency. Such interactions are often missed by overly efficient RNAi reagents. We therefore developed an assay that measures viability over a range of knockdown efficiency within a cell population. This method, called Variable Dose Analysis (VDA), is highly sensitive to viability phenotypes and reproducibly detects SS/L interactions. We applied the VDA method to search for SS/L interactions with
Sigma-Aldrich
MISSION® esiRNA, targeting human ARCN1
Sigma-Aldrich
MISSION® esiRNA, targeting human NRGN