MilliporeSigma
  • In situ fabrication of cobalt nanoflowers on sulfonated and fluorinated poly (arylene ether ketone-benzimidazole) template film for the electrocatalytic oxidation of glucose.
In situ fabrication of cobalt nanoflowers on sulfonated and fluorinated poly (arylene ether ketone-benzimidazole) template film for the electrocatalytic oxidation of glucose.
Talanta (2017-11-16)
Tengfei Wang, Lingling Xi, Jianli Wang
ABSTRACT
Using sulfonated and fluorinated poly (arylene ether ketone) comprising functional strong coordination group benzimidazole (SPAEK-F-BI) as a template film, a novel fabrication method of cobalt nanoflowers (CoNFs) and non-enzymatic glucose electrochemical sensor was developed in this work. After the precursors Co
Sigma-Aldrich
4,4′-(Hexafluoroisopropylidene)diphenol, 97%