MilliporeSigma
  • Biomarker analysis of hemoglobin adducts of acrylamide and glycidamide enantiomers for mid-term internal exposure assessment by isotope dilution ultra-high performance liquid chromatography tandem mass spectrometry.
Biomarker analysis of hemoglobin adducts of acrylamide and glycidamide enantiomers for mid-term internal exposure assessment by isotope dilution ultra-high performance liquid chromatography tandem mass spectrometry.
Talanta (2017-11-16)
Yu Zhang, Qiao Wang, Gong Zhang, Wei Jia, Yiping Ren, Yongning Wu
ABSTRACT
Hemoglobin (Hb) adducts of acrylamide (AA) and its oxidative metabolite glycidamide (GA) are important biomarkers for evaluating the mid-term exposure of acrylamide toxicity in vivo. Taking pentafluoro-2-methylphenyl isothiocyanates of N-(2-carbamoylethyl)valine (AAVal-PFPTH) and N-(2-carbamoyl-2-hydroxyethy)valine (GAVal-PFPTH) as target analytes, we developed an isotope dilution ultra-high performance liquid chromatograph tandem mass spectrometry (UHPLC-MS/MS) method for the simultaneous determination of AA and GA hemoglobin (Hb) adducts under the electroscopy ionization negative (ESI‾) mode in the present work. Among them, the enantiomer pair of GA-Hb adducts was firstly identified and successfully separated at baseline level. The method achieved high sensitivity with the LOD and LOQ ranging 1.43-5.05pmol/g Hb and 4.78-16.82pmol/g Hb, respectively. The recovery rates with low, intermediate and high spiking levels were calculated as 97.0-105.2%, 97.4-106.4% and 100.3-111.2%, respectively. Acceptable within-laboratory reproducibility (RSD < 13.7%) substantially supported the robustness of current UHPLC-MS/MS method, which was successfully applied to measure the hemoglobin adducts of acrylamide and glycidamide enantiomers in blood of both rats and humans. A linear exposure assessment model was developed for estimating the daily exposure to acrylamide in humans via considering acrylamide hemoglobin adducts as variables, indicating a novel connect between biomarker-based internal exposure and dietary-based external exposure. Overall, the present instrumental analysis and related internal exposure assessment model provide a substantially methodological support for profiling the internal biological exposure and estimating the external dietary exposure to acrylamide.
Sigma-Aldrich
Pentafluorophenyl isothiocyanate, ≥97.0% (GC)
Supelco
Glycidamide, analytical standard